EMAIL THIS PAGE TO A FRIEND
Indian journal of medical microbiology
Emergence of optochin resistance among S. pneumoniae strains colonizing healthy children in Abidjan.
PMID 20061777
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E8253
Ethylhydrocupreine, analytical standard
C21H28N2O2
E9876
Ethylhydrocupreine hydrochloride, analytical standard
C21H28N2O2 · HCl