EMAIL THIS PAGE TO A FRIEND
TheScientificWorldJournal
Polycyclic aromatic hydrocarbon concentration levels on the Korean peninsula between 2006 and 2008.
PMID 20062948
Abstract
Concentrations of seven polycyclic aromatic hydrocarbon (PAH) compounds - benzo(a)anthracene (BaA), chrysene (CHRY), benzo(b)fluoranthene (BbF), benzo(k)fluoranthene (BkF), dibenz(a,h)anthracene (DahA), indeno(1,2,3-cd)pyrene (I123P), and benzo(a)pyrene (BaP) - in air were measured as the sum of gas and particle fractions at 32 monitoring stations dispersed across Korea during a 2-year period (February 2006 to January 2008). The data sets were collected at intervals of 1 day (24 h) per month from each monitoring station. According to our analysis, the spatial distribution of PAH is distinguished by manmade activities between different land use types. Evaluation of total PAH (T-PAH) concentration levels, which were derived by summing up all individual compounds, revealed that the T-PAH value varied on the order of commercial (4.85 + or - 4.40 ng m(-3)) rural (4.42 + or - 2.73 ng m(-3)), industrial (4.27 + or - 1.79 ng m(-3)), greenland (3.09 + or - 3.86 ng m(-3)), and background (2.60 + or - 2.54 ng m(-3)) areas. The PAH values, when compared across seasons, tend to peak consistently during the winter (or spring) due to the active consumption of fossil fuels. The overall results of this study confirm that the pollution status of PAH compounds are clearly discernible not only between areas with different levels of anthropogenic activities, but also between periods with changes in environmental conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B2209
Benz[a]anthracene, 99%
C18H12
48563
Benz[a]anthracene, analytical standard
C18H12
BCR271
Benz[a]anthracene, BCR® certified Reference Material
C18H12
75451
Benz[a]anthracene, certified reference material, TraceCERT®
C18H12
36956
Benz[a]anthracene solution, 100 ng/μL in cyclohexane, analytical standard
C18H12
48651
Benz[a]anthracene solution, certified reference material, 200 μg/mL in methylene chloride
C18H12
40070-U
Benz[a]anthracene solution, certified reference material, 1000 μg/mL in methanol
C18H12
48490
Benzo[b]fluoranthene, analytical standard
C20H12
BCR047
Benzo[b]fluoranthene, BCR® certified Reference Material
C20H12
30958
Benzo[b]fluoranthene, certified reference material, TraceCERT®
C20H12
SCB-002
Benzo[b]fluoranthene, vial of 1 g, analytical standard
C20H12
275336
Benzo[b]fluoranthene, 98%
C20H12
36955
Benzo[b]fluoranthene solution, 100 μg/mL in cyclohexane, analytical standard
C20H12
48637
Benzo[b]fluoranthene solution, certified reference material, 200 μg/mL in methanol
C20H12
40072
Benzo[b]fluoranthene solution, certified reference material, 1000 μg/mL in acetone
C20H12
392251
Benzo[k]fluoranthene, for fluorescence, ≥99%
C20H12
BCR048R
Benzo[k]fluoranthene, BCR® certified Reference Material
C20H12
03323
Benzo[k]fluoranthene, certified reference material, TraceCERT®
C20H12
48492
Benzo[k]fluoranthene, analytical standard
C20H12
48668
Benzo[k]fluoranthene solution, certified reference material, 200 μg/mL in methylene chloride
C20H12
40073
Benzo[k]fluoranthene solution, certified reference material, 1000 μg/mL in acetone
C20H12
49476-U
Benzo(k)fluoranthene solution, 100 μg/mL in methylene chloride, analytical standard
C20H12
35754
Chrysene, analytical standard
C18H12
48565-U
Chrysene, analytical standard
C18H12
BCR269
Chrysene, BCR® certified Reference Material
C18H12
94035
Chrysene, certified reference material, TraceCERT®
C18H12
49478-U Chrysene solution, 100 μg/mL in methylene chloride, analytical standard
40074 Chrysene solution, certified reference material, 1000 μg/mL in acetone
BCR138
Dibenz[a,h]anthracene, BCR® certified Reference Material
C22H14
91861
Dibenz[a,h]anthracene, certified reference material, TraceCERT®
C22H14