EMAIL THIS PAGE TO A FRIEND
Journal of enzyme inhibition and medicinal chemistry
Characterisation of hydrazides and hydrazine derivatives as novel aspartic protease inhibitors.
PMID 20063996
Abstract
Virtual screening of an in-house virtual library of synthetic compounds using FlexX, followed by enzyme inhibition, identified hydrazide and hydrazine derivatives as novel aspartic protease inhibitors. These compounds inhibited human cathepsin D and Plasmodium falciparum plasmepsin-II with low micromolar concentrations (IC(50) = 1-2.5 microM). Modelling studies with plasmepsin-II predicted binding of ligands at the centre of the extended substrate-binding cleft, where hydrazide/hydrazine parts of the inhibitors acted as the transition state mimic by forming electrostatic interactions with catalytic aspartates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
442232-U
1,2-Diphenylhydrazine, analytical standard, ampule of 100 mg
C12H12N2
126721
Hydrazobenzene
C12H12N2