EMAIL THIS PAGE TO A FRIEND
Arthritis research & therapy
The effect of the pro-inflammatory cytokine tumor necrosis factor-alpha on human joint capsule myofibroblasts.
PMID 20064200
Abstract
Previous studies have shown that the number of myoblastically differentiated fibroblasts known as myofibroblasts (MFs) is significantly increased in stiff joint capsules, indicating their crucial role in the pathogenesis of post-traumatic joint stiffness. Although the mode of MFs' function has been well defined for different diseases associated with tissue fibrosis, the underlying mechanisms of their regulation in the pathogenesis of post-traumatic joint capsule contracture are largely unknown. In this study, we examined the impact of the pro-inflammatory cytokine tumor necrosis factor-alpha (TNF-alpha) on cellular functions of human joint capsule MFs. MFs were challenged with different concentrations of TNF-alpha with or without both its specifically inactivating antibody infliximab (IFX) and cyclooxygenase-2 (COX2) inhibitor diclofenac. Cell proliferation, gene expression of both alpha-smooth muscle actin (alpha-SMA) and collagen type I, the synthesis of prostaglandin derivates E2, F1A, and F2A, as well as the ability to contract the extracellular matrix were assayed in monolayers and in a three-dimensional collagen gel contraction model. The alpha-SMA and COX2 protein expressions were evaluated by immunofluorescence staining and Western blot analysis. The results indicate that TNF-alpha promotes cell viability and proliferation of MFs, but significantly inhibits the contraction of the extracellular matrix in a dose-dependent manner. This effect was associated with downregulation of alpha-SMA and collagen type I by TNF-alpha application. Furthermore, we found a significant time-dependent upregulation of prostaglandin E2 synthesis upon TNF-alpha treatment. The effect of TNF-alpha on COX2-positive MFs could be specifically prevented by IFX and partially reduced by the COX2 inhibitor diclofenac. Our results provide evidence that TNF-alpha specifically modulates the function of MFs through regulation of prostaglandin E2 synthesis and therefore may play a crucial role in the pathogenesis of joint capsule contractures.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P5765
Prostaglandin F, synthetic
C20H36O5