EMAIL THIS PAGE TO A FRIEND
The American journal of tropical medicine and hygiene
Sodium dichloroisocyanurate tablets for routine treatment of household drinking water in periurban Ghana: a randomized controlled trial.
PMID 20064989
Abstract
We conducted a randomized, placebo-controlled, triple-blinded trial to determine the health impact of daily use of sodium dichloroisocyanurate (NaDCC) tablets for household drinking water treatment in periurban Ghana. We randomized 240 households (3,240 individuals) to receive either NaDCC or placebo tablets. All households received a 20-liter safe water storage vvessel. Over 12 weeks, 446 diarrhea episodes (2.2%) occurred in intervention and 404 (2.0%) in control households (P = 0.38). Residual free chlorine levels indicated appropriate tablet use. Escherichia coli was found in stored water at baseline in 96% of intervention and 88% of control households and at final evaluation in 8% of intervention and 54% of control households (P = 0.002). NaDCC use did not prevent diarrhea but improved water quality. Diarrhea rates were low and water quality improved in both groups. Safe water storage vessels may have been protective. A follow-up health impact study of NaDCC tablets is warranted.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
218928
Sodium dichloroisocyanurate, 96%
C3Cl2N3NaO3
35915
Sodium dichloroisocyanurate dihydrate, ≥98.0% (AT)
C3Cl2N3NaO3 · 2H2O