EMAIL THIS PAGE TO A FRIEND
Organic letters
Construction of a library of rhodol fluorophores for developing new fluorescent probes.
PMID 20067265
Abstract
A highly efficient and concise synthetic scheme for rhodol fluorophores is developed with palladium-catalyzed amination reaction as the key step. This new synthetic route is utilized to construct a rhodol library, potentially useful for the design of novel fluorescent probes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
67806
Rhodol, ≥85% (HPCE)
C20H13NO4