EMAIL THIS PAGE TO A FRIEND
Journal of economic entomology
Ability of two natural products, nootkatone and carvacrol, to suppress Ixodes scapularis and Amblyomma americanum (Acari: Ixodidae) in a Lyme disease endemic area of New Jersey.
PMID 20069863
Abstract
We evaluated the ability of the natural, plant-derived acaricides nootkatone and carvacrol to suppress Ixodes scapularis Say and Amblyomma americanum (L.) (Acari: Ixodidae). Aqueous formulations of 1 and 5% nootkatone applied by backpack sprayer to the forest litter layer completely suppressed I. scapularis nymphs through 2 d. Thereafter, the level of reduction gradually declined to < or =50% at 28 d postapplication. Against A. americanum nymphs, 1% nootkatone was less effective, but at a 5% concentration, the level of control was similar or greater to that observed with I. scapularis through 21 d postapplication. Initial applications of 0.05% carvacrol were ineffective, but a 5% carvacrol formulation completely suppressed nymphs of both species through 2 d and resulted in significant reduction in I. scapularis and A. americanum nymphs through 28 and 14 d postapplication, respectively. Backpack sprayer applications of 5% nootkatone to the shrub and litter layers resulted in 100% control of I. scapularis adults through 6 d, but the level of reduction declined to 71.5% at 28 d postapplication. By contrast, high-pressure applications of 2% nootkatone to the litter layer resulted in 96.2-100% suppression of both I. scapularis and A. americanum nymphs through 42 d, whereas much lower control was obtained from the same formulation applied by backpack sprayer. Backpack sprayer application of a 3.1% nootkatone nanoemulsion resulted in 97.5-98.9 and 99.3-100% reduction in I. scapularis and A. americanum nymphs, respectively, at 1 d postapplication. Between 7 d and 35 d postapplication, the level of control varied between 57.1% and 92.5% for I. scapularis and between 78.5 and 97.1% for A. americanum nymphs. The ability of natural products to quickly suppress and maintain significant control of populations of these medically important ticks at relatively low concentrations may represent a future alternative to the use of conventional synthetic acaricides.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
74437
(+)-Nootkatone, ≥99.0% (GC)
C15H22O
93785
(+)-Nootkatone, technical, ≥85% (GC)
C15H22O