EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Chemical constituents of roots of Boehmeria nivea].
PMID 20069903
Abstract
To investigate the chemical constituents from roots of Boehmeria nivea. The constituents were isolated by repeated column chromatography and preparative liquid chromatography; and their structures were elucidated by chemical properties and spectroscopic analyses. Seven compounds were isolated and their structures were identified as tormentic acid (1), hederagenin (2), maslinic acid (3), 2alpha-hydroxyursolic acid (4), trans-p-hydroxycinamic acid (5), 2,4,4'-trihydroxychalcone (6), rutin (7). Compounds 1-6 were obtained from this genus for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H3916
Hederagenin, ≥97% (HPLC)
C30H48O4