EMAIL THIS PAGE TO A FRIEND
The Journal of neuroscience : the official journal of the Society for Neuroscience
Collapsin response mediator protein 4a (CRMP4a) is upregulated in motoneurons of mutant SOD1 mice and can trigger motoneuron axonal degeneration and cell death.
PMID 20071543
Abstract
Embryonic motoneurons from mutant SOD1 (mSOD1) mouse models of amyotrophic lateral sclerosis (ALS), but not wild-type motoneurons, can be triggered to die by exposure to nitric oxide (NO), leading to activation of a motoneuron-specific signaling pathway downstream of the death receptor Fas/CD95. To identify effectors of mSOD1-dependent cell death, we performed a proteomic analysis. Treatment of cultured mSOD1 motoneurons with NO led to a 2.5-fold increase in levels of collapsin response mediator protein 4a (CRMP4a). In vivo, the percentage of mSOD1 lumbar motoneurons expressing CRMP4 in mSOD1 mice increased progressively from presymptomatic to early-onset stages, reaching a maximum of 25%. Forced adeno-associated virus (AAV)-mediated expression of CRMP4a in wild-type motoneurons in vitro triggered a process of axonal degeneration and cell death affecting 60% of motoneurons, whereas silencing of CRMP4a in mSOD1 motoneurons protected them from NO-induced death. In vivo, AAV-mediated overexpression of CRMP4a but not CRMP2 led to the death of 30% of the lumbar motoneurons and an 18% increase in denervation of neuromuscular junctions in the gastrocnemius muscle. Our data identify CRMP4a as a potential early effector in the neurodegenerative process in ALS.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MAB377
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse