EMAIL THIS PAGE TO A FRIEND
Small (Weinheim an der Bergstrasse, Germany)
Reduced graphene oxide-templated photochemical synthesis and in situ assembly of Au nanodots to orderly patterned Au nanodot chains.
PMID 20077425
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O1858
1-Octadecanethiol, 98%
C18H38S
74731
1-Octadecanethiol, purum, ≥95.0% (GC)
C18H38S