EMAIL THIS PAGE TO A FRIEND
Thrombosis research
Contribution of surface localization to platelet activation by CD154 antibodies and introduction of a novel approach for studying immune complex activity.
PMID 20083297