EMAIL THIS PAGE TO A FRIEND
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine
Effects of structural differences between radioiodine-labeled 1-(2'-fluoro-2'-deoxy-D-arabinofuranosyl)-5-iodouracil (FIAU) and 1-(2'-fluoro-2'-deoxy-d-ribofuranosyl)-5-iodouracil (FIRU) on HSV1-TK reporter gene imaging.
PMID 20083410
Abstract
The goal of this investigation was to evaluate the effects of structural differences between FIAU and FIRU on their ability to serve as a potential tracer for reporter gene imaging. To examine the characteristics of different configurations of FIAU and FIRU, a series of evaluations were done on HSV1-TK gene-expressing cells and on mice with HSV1-TK gene-expressing tumor. The results showed that, as an imaging agent for HSV1-TK-expressing cells, radiolabeled FIAU was more efficient for in vivo imaging than FIRU.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0632
Fialuridine, ≥98% (HPLC)
C9H10FIN2O5