EMAIL THIS PAGE TO A FRIEND
Journal of fluorescence
The role of pyranine in characterization of PAAm-κC composites by using fluorescence technique.
PMID 20084435
Abstract
Polyacrylamide (PAAm) doped by κ-carrageenan (κC) gels were prepared with various amounts of κC varying in the range between 0 wt.% and 3 wt.%. Steady-state fluorescence (SSF) technique was employed for studying sol-gel transition and swelling of PAAm-κC composite gels which were prepared by free-radical crosslinking copolymerization. Pyranine was introduced as a fluorescence probe. Pyranine molecules start to bind to acrylamide polymer chains upon the initiation of the polymerization, thus the spectra of the bonded pyranines shift to the shorter wavelengths. Fluorescence spectra from the bonded pyranines allow one to monitor the sol-gel transition and to test the universality of the sol-gel transition as a function of some kinetic parameters like polymer concentration. Observations around the gel point, t ( c ) for PAAm-κC composite gels showed that the gel fraction exponent β obeyed the percolation result for low κC (<2.0 wt. %) however classical results were produced at higher κC (>2.0 wt.%). On the other hand, fluorescence intensity of pyranine was measured during in situ swelling process at various amounts of κC and it was observed that fluorescence intensity values decreased as swelling is proceeded. Li-Tanaka equation was used to determine the swelling time constants, τ and cooperative diffusion coefficients, D.