EMAIL THIS PAGE TO A FRIEND
The journal of physical chemistry. A
Structure of zirconocene complexes relevant for olefin catalysis: infrared fingerprint of the Zr(C(5)H(5))(2)(OH)(CH(3)CN)(+) cation in the gas phase.
PMID 20085359
Abstract
Cationic zirconocene complexes are active species in Ziegler-Natta catalysis for olefin polymerization. Their structure and metal-ligand bond strength strongly influence their activity. In the present work, the infrared multiphoton dissociation (IRMPD) spectrum of mass selected Zr(C(5)H(5))(2)(OH)(CH(3)CN)(+) cations was obtained in the 300-1500 cm(-1) fingerprint range by coupling a Fourier-transform ion cyclotron resonance (FT-ICR) mass spectrometer equipped with an electrospray ionization (ESI) source and the infrared free electron laser (IR-FEL) at the Centre Laser Infrarouge d'Orsay (CLIO). The experimental efforts are complemented by quantum chemical calculations at the MP2 and B3LYP levels using the 6-311G* basis set. Vibrational assignments of transitions observed in the IRMPD spectra to modes of the Zr-O-H, C(5)H(5), and CH(3)CN moieties are based on comparison to calculated linear absorption spectra. Both the experimental data and the calculations provide unprecedented information about structure, metal-ligand bonding, charge distribution, and binding energy of the complex.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
196215
Bis(cyclopentadienyl)zirconium(IV) dichloride, ≥98%
C10H10Cl2Zr