EMAIL THIS PAGE TO A FRIEND
International journal of toxicology
Final report on the safety assessment of Basic Violet 1, Basic Violet 3, and Basic Violet 4.
PMID 20086192
Abstract
Basic Violet 3, Basic Violet 1, and Basic Violet 4 are triphenylmethane dyes that function as direct (nonoxidative) hair colorants. No current uses or use concentrations in cosmetics are reported. The term Gentian Violet is used synonymously with Basic Violet 1 and Basic Violet 3, although the chemical structures of these 2 dyes are not the same. The Cosmetic Ingredient Review Expert Panel noted that Basic Violet 1, 3, and 4 contain quaternary ammonium ions, and therefore the rate of penetration across the epidermis is expected to be slow. The panel concluded that because of the carcinogenic potential of these dyes, insufficient data exist to support the safety of Basic Violet 1, 3, and 4 in cosmetic formulation. Dermal absorption data and a risk assessment are needed to complete this safety assessment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
228842
Ethyl Violet, cationic triarylmethane dye
C31H42ClN3