EMAIL THIS PAGE TO A FRIEND
Journal of virology
E1A interacts with two opposing transcriptional pathways to induce quiescent cells into S phase.
PMID 20089639
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
05-257
Anti-p300 CT Antibody, clone RW128, clone RW128, Upstate®, from mouse