EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
A synthesis of oligomeric alpha-hydroxy phenylphosphinates and a study of the conformational preferences of the dimers.
PMID 20090976
Abstract
A general method for the synthesis of a novel class of oligomers, comprising alpha-hydroxy phenylphosphinic acid building blocks, is reported. A series of dimeric alpha-hydroxy phenylphosphinates are analyzed by a combination of NMR spectroscopy, X-ray crystallography and computational methods.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P29006
Phenylphosphonic acid, 98%
C6H7O3P