EMAIL THIS PAGE TO A FRIEND
Organic letters
Synthesis of benzoxazolones from nitroarenes or aryl halides.
PMID 20092263
Abstract
A simple and effective method for the preparation of benzoxazolones from nitroarenes or aryl halides is described. Partial reduction of a nitro group in the presence of a chloroformate followed by a microwave-assisted rearrangement/ring closure sequence provides a convenient and practical procedure to prepare this important pharmacophore. Rearrangement precursors were also accessed from aryl halides through transition-metal-catalyzed coupling.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
157058
2-Benzoxazolinone, 98%
C7H5NO2