EMAIL THIS PAGE TO A FRIEND
Bioconjugate chemistry
Synthesis of conjugates of 6-aminophenanthridine and guanabenz, two structurally unrelated prion inhibitors, for the determination of their cellular targets by affinity chromatography.
PMID 20092293
Abstract
The synthesis of affinity matrices for 6-aminophenanthridine (6AP) and 2,6-dichlorobenzylidenaminoguanidine (Guanabenz, GA), two unrelated prion inhibitors, is described. In both cases, the same simple spacer, epsilon-aminocaproylaminopentanol, was introduced by a Mitsunobu reaction and the choice of the anchoring position of the linker was determined by the study of the residual antiprion activity of the corresponding 6AP or GA conjugates. Very recently, these two affinity matrices were used for chromatography assays leading to the identification of ribosome (via the rRNA) as a common target of these two antiprion drugs. Here, we show, using competition experiments with Quinacrine (QC) and Chlorpromazine (CPZ), two other antiprion drugs, that QC, but not CPZ, may also directly target the rRNA.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G110
Guanabenz acetate salt, powder
C8H8Cl2N4 · C2H4O2