EMAIL THIS PAGE TO A FRIEND
Chemical research in toxicology
Cytochrome P450-mediated epoxidation of 2-aminothiazole-based AKT inhibitors: identification of novel GSH adducts and reduction of metabolic activation through structural changes guided by in silico and in vitro screening.
PMID 20095585
Abstract
A 2-aminothiazole derivative 1 was developed as a potential inhibitor of the oncology target AKT, a serine/threonine kinase. When incubated in rat and human liver microsomes in the presence of NADPH, 1 underwent significant metabolic activation on its 2-aminothiazole ring, leading to substantial covalent protein binding. Upon addition of glutathione, covalent binding was reduced significantly, and multiple glutathione adducts were detected. Novel metabolites from the in vitro incubates were characterized by LC-MS and NMR to discern the mechanism of bioactivation. An in silico model was developed based on the proposed mechanism and was employed to predict bioactivation in 23 structural analogues. The predictions were confirmed empirically for the bioactivation liability, in vitro, by LC-MS methods screening for glutathione incorporation. New compounds were identified with a low propensity for bioactivation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
123129
2-Aminothiazole, 97%
C3H4N2S