EMAIL THIS PAGE TO A FRIEND
Clinical toxicology (Philadelphia, Pa.)
A modified low-cost colorimetric method for paracetamol (acetaminophen) measurement in plasma.
PMID 20095813
Abstract
Despite a significant increase in the number of patients with paracetamol poisoning in the developing world, plasma paracetamol assays are not widely available. The purpose of this study was to assess a low-cost modified colorimetric paracetamol assay that has the potential to be performed in small laboratories with restricted resources. The paracetamol assay used in this study was based on the Glynn and Kendal colorimetric method with a few modifications to decrease the production of nitrous gas and thereby reduce infrastructure costs. Preliminary validation studies were performed using spiked aqueous samples with known concentrations of paracetamol. Subsequently, the results from the colorimetric method for 114 stored clinical samples from patients with paracetamol poisoning were compared with those from the current gold-standard high-performance liquid chromatography method. A prospective survey, assessing the clinical use of the paracetamol assay, was performed on all patients with paracetamol poisoning attending the Peradeniya General Hospital, Sri Lanka, over a 10-month period. The recovery study showed an excellent correlation (r(2) > 0.998) for paracetamol concentrations from 25 to 400 mg/L. The final yellow color was stable for at least 10 min at room temperature. There was also excellent correlation with the high-performance liquid chromatography method (r(2) = 0.9758). In the clinical cohort study, use of the antidote N-acetylcysteine was avoided in over a third of patients who had the plasma paracetamol concentration measured. The cost of consumables used per assay was $0.50 (US). This colorimetric paracetamol assay is reliable and accurate and can be performed rapidly, easily, and economically. Use of this assay in resource-poor clinical settings has the potential to have a significant clinical and economic impact on the management of paracetamol poisoning.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
383120
Sulfamic acid, ACS reagent, 99.3%
H3NO3S