EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Synthesis and structure-activity relationships of a series of 3-aryl-4-isoxazolecarboxamides as a new class of TGR5 agonists.
PMID 20097073
Abstract
A series of 3-aryl-4-isoxazolecarboxamides identified from a high-throughput screening campaign as novel, potent agonists of the human TGR5 G-protein-coupled receptor is described. Many analogues were readily accessible via solution-phase synthesis which resulted in the rapid identification of key structure-activity relationships (SAR), and the discovery of potent exemplars (up to pEC50=9). Details of the SAR and optimization of this series are presented herein.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
210358
4-Chloro-N-methylaniline, 97%
C7H8ClN