EMAIL THIS PAGE TO A FRIEND
Clinical and vaccine immunology : CVI
IgA antibody response of swine to foot-and-mouth disease virus infection and vaccination.
PMID 20107003
Abstract
Foot-and-mouth disease virus (FMDV) continues to be a significant economic problem worldwide. Control of the disease involves the use of killed-virus vaccines, a control measure developed decades ago. After natural infection, the primary site of replication of FMDV is the pharyngeal area, suggesting that a mucosal immune response is the most effective. Humoral immunity to killed-virus vaccination induces antibodies that can prevent the clinical disease but not local infection. Determining whether infection or vaccination stimulates IgA-mediated local immunity depends on the method of analysis. Different assays have been described to analyze the quality of antibody responses of cattle and swine to FMDV, including indirect double-antibody sandwich enzyme-linked immunosorbent assay (IDAS-ELISA) and antibody capture assay-ELISA (ACA-ELISA). We tested these assays on swine and show that vaccinated animals had FMDV-specific IgM and IgG but no IgA in either serum or saliva. After the infection, both assays detected FMDV-specific IgM, IgG, and IgA in serum. Notably, serum IgA was more readily detected using the ACA-ELISA, whereas IgA was not detected in saliva with this assay. FMDV-specific IgA antibodies were detected in saliva samples using the IDAS-ELISA. These data show that parenterally administered, killed-virus vaccine does not induce a mucosal antibody response to FMDV and illuminates limitations and appropriate applications of the two ELISAs used to measure FMDV-specific responses. Further, the presence of the IgA antivirus in serum correlates with the presence of such antibodies in saliva.