EMAIL THIS PAGE TO A FRIEND
Journal of chromatographic science
Simultaneous determination of ezetimibe and simvastatin in pharmaceutical preparations by MEKC.
PMID 20109284
Abstract
A micellar electrokinetic capillary chromatography method was developed and validated for the simultaneous determination of ezetimibe and simvastatin in pharmaceutical preparations. The influence of buffer concentration, buffer pH, sodium dodecyl sulphate (SDS) concentration, organic modifier, capillary temperature, applied voltage, and injection time was investigated, and the method validation studies were performed. The optimum separation for these analytes was achieved in less than 10 min at 30 degrees C with a fused-silica capillary column (56 cm x 50 microm i.d.) and a 25mM borate buffer at pH 9.0 containing 25mM SDS and 10% (v/v) acetonitrile. The samples were injected hydrodynamically for 3 s at 50 mbar, and the applied voltage was +30.0 kV. Detection wavelength was set at 238 nm. Diflunisal was used as internal standard. The method was suitably validated with respect to stability, specificity, linearity, limits of detection and quantification, accuracy, precision, and robustness. The limits of detection and quantification were 1.0 and 2.0 microg/mL for both ezetimibe and simvastatin, respectively. The method developed was successfully applied to the simultaneous determination of ezetimibe and simvastatin in pharmaceutical preparations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3281
Diflunisal, analytical standard
C13H8F2O3