EMAIL THIS PAGE TO A FRIEND
Planta medica
Lapachol induces clastogenic effects in rats.
PMID 20112181
Abstract
Lapachol is a naturally occurring naphthoquinone derivative found in the heartwood of several plants, particularly those of the genus Tabebuia (Bignoneaceae). Despite its use as a therapeutic product with anti-inflammatory, analgesic, antipsoriatic, trypanocidal effects, among others, its in vivo mutagenic potential has still not been investigated. This paper reports the effects after a single oral administration of lapachol in the in vivo micronucleus (MN) and chromosome aberration (CA) assays. Both assays were performed using bone marrow cells from male Wistar rats. The animals were treated by oral gavage with hydroalcoholic solutions of lapachol at the doses of 122, 244 and 365 mg/kg, chosen on the basis of the LD(50) in male rats. The results show that the higher administered lapachol dose induced a significant increase in the frequency of micronucleated polychromatic erythrocytes (MNPCE) and CAs in rat bone marrow cells, indicating that lapachol shows clastogenic effects under the experimental conditions used.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
142905
Lapachol, 98%
C15H14O3