EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Pyridine-functionalized MCM-41 as an efficient and recoverable catalyst for the synthesis of pyran annulated heterocyclic systems.
PMID 20118595
Abstract
Pyridine-functionalized MCM-41 catalyzed reactions between tetracyanoethylene and various activated CH-acid compounds are described. These reactions afford the corresponding pyran annulated heterocyclic ring systems in high yields at room temperature within a few minutes. The work-up procedure is very simple and the products do not require further purification. The catalyst can be recycled and reused for several times without observable loss of performance.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T8809
Tetracyanoethylene, 98%
C6N4