EMAIL THIS PAGE TO A FRIEND
The journal of physical chemistry. B
Monolayer and Brewster angle microscopy studies of poly(methyl methacrylate)-monopalmitin mixed systems at the air-water interface.
PMID 20121203
Abstract
Mixed monolayers of poly(methyl metacrylate) (PMMA) and monopalmitin (Mp) were used for the study of their interactions. A thorough analysis of surface pressure (pi)-area (A) isotherms with the Langmuir monolayer technique, complemented with Brewster angle microscopy (BAM) images was performed. Mixed films show two phase transitions at a surface pressure of 14.5 mN/m and at 20-21 mN/m, respectively. Moreover, mixed monolayers show two well-defined collapses: one, corresponding to the lipid (at surface pressures of 50-51 mN/m) and the another one, ascribed to the PMMA, at surface pressure values of 57-58 mN/m. When the mean molecular areas of the mixed films (A(1,2)) were plotted versus film composition (X(1) or X(2)), positive deviations from the ideal behavior were observed at surface pressures below 15 mN/m, which were mainly attributed to a change in the conformation of the PMMA molecules at the surface. However, at higher surface pressures, the areas per monomer unit of the mixed monolayers obey the additivity rule, attributed to the fact that the film components form an immiscible system in these conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M1640
DL-α-Palmitin, ≥99%
C19H38O4