EMAIL THIS PAGE TO A FRIEND
Environmental health perspectives
Association of biomarkers of systemic inflammation with organic components and source tracers in quasi-ultrafine particles.
PMID 20123637
Abstract
Evidence is needed regarding the air pollutant components and their sources responsible for associations between particle mass concentrations and human cardiovascular outcomes. We previously found associations between circulating biomarkers of inflammation and mass concentrations of quasi-ultrafine particles