EMAIL THIS PAGE TO A FRIEND
Developmental Dynamics
The relationship of the birth date of rat sympathetic neurons to the target they innervate.
PMID 20131352
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB1583 Anti-Neuropeptide Y Antibody, serum, clone, Chemicon®