EMAIL THIS PAGE TO A FRIEND
Toxicology and Applied Pharmacology
The effects of 3,4-methylenedioxymethamphetamine (MDMA) on nicotinic receptors: intracellular calcium increase, calpain/caspase 3 activation, and functional upregulation.
PMID 20132834