EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Simultaneous measurement of S-warfarin, R-warfarin, S-7-hydroxywarfarin and R-7-hydroxywarfarin in human plasma by liquid chromatography-tandem mass spectrometry.
PMID 20133101
Abstract
Clinically used anticoagulant warfarin is usually available as a racemic mixture of S- and R-warfarin that are both metabolized mainly by cytochrome P450 isoenzymes. Determination of warfarin enantiomers and their enantiomeric 7-hydroxywarfarin (7-OH-warfarin) metabolites in the human plasma sample allows probing of cytochrome P450 isoenzyme activities and detection of ingestion of warfarin-containing products for the investigation of unexplained bleeding. The present study aims to develop a sensitive and specific liquid chromatography-tandem mass spectrometry (LC-MS/MS) method for simultaneous detection of S-warfarin, R-warfarin, S-7-OH-warfarin and R-7-OH-warfarin in human plasma. Plasma samples were extracted with mixed-mode cation-exchange (MCX) cartridge with recoveries of greater than 87.0% for all four analytes. The selectivity of 7-OH-warfarin from other monohydroxylated warfarin metabolites such as 4'-, 6-, 8- and 10-hydroxywarfarins using a Chirobiotic V chiral column and multiple reaction monitoring (MRM) was addressed. The developed LC-MS/MS method is simple, specific and has been fully validated with satisfactory accuracy and adequate reproducibility with limit of quantification (LOQ) of 5 ng/ml for all four analytes. The method was successfully applied to analyze the steady state plasma concentrations of the four analytes in 30 patients.