EMAIL THIS PAGE TO A FRIEND
Nature protocols
Experimental acute biliary pancreatitis induced by retrograde infusion of bile acids into the mouse pancreatic duct.
PMID 20134432
Abstract
Mechanistic studies of acute pancreatitis require animal models because clinical material is generally not available during the early phases of the disease. Here we describe a protocol to induce biliary pancreatitis by retrogradely infusing bile acids into the pancreatic duct of anesthetized mice. The resulting model replicates events believed to be responsible for the onset of clinical biliary (i.e., gallstone) pancreatitis and creates highly reproducible pancreatitis with a severity that depends on the concentration of infused bile acid. Pancreatitis reaches its maximal level of severity within 24 h of induction, and it resolves over the subsequent week. This protocol enables the investigator to use genetically modified strains of mice, and it requires only relatively simple and easily learned techniques of small animal surgery. With practice and gentle technique, the surgery (from induction of anesthesia to completion of the infusion) can be completed within 25 min per animal.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T7515
Sodium taurolithocholate
C26H44NNaO5S