EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Nucleophilic heterocyclic carbene as a novel catalyst for cyclopropanation of cyano acrylates.
PMID 20135050
Abstract
Nucleophilic heterocyclic carbenes (NHCs) have been used for the first time as catalysts in the cyclopropanation of ethyl cyanocinnamates with phenacyl bromide by Michael-initiated ring-closure (MIRC).
Related Materials