EMAIL THIS PAGE TO A FRIEND
Phytochemistry
Constituents and secondary metabolite natural products in fresh and deteriorated cassava roots.
PMID 20137795
Abstract
A phytochemical analysis of cassava (Manihot esculenta Crantz) fresh roots and roots suffering from post-harvest physiological deterioration (PPD) has been carried out. The first isolation and identification of galactosyl diacylglycerides from fresh cassava roots is reported, as well as beta-carotene, linamarin, and beta-sitosterol glucopyranoside. The hydroxycoumarin scopoletin and its glucoside scopolin were identified from cassava roots during PPD, as well as trace quantities of esculetin and its glucoside esculin. There is no isoscopoletin in cassava roots during PPD.
Related Materials