EMAIL THIS PAGE TO A FRIEND
Journal of colloid and interface science
Superhydrophobic PLA fabrics prepared by UV photo-grafting of hydrophobic silica particles possessing vinyl groups.
PMID 20138632
Abstract
Superhydrophobic poly(lactic acid) (PLA) fabrics are prepared by UV photo-grafting of hydrophobic silica particles possessing vinyl functional groups on the surfaces, which is a novel one-step process to provide surface with roughness as well as hydrophobicity simultaneously. For this purpose, hydrophobic silica particles with vinyl groups and average diameter of 1.51+/-0.05 microm are synthesized via a sol-gel process. The silica particles possessing vinyl groups are found to be effectively immobilized on PLA fabrics via UV photo-grafting reaction. The water contact angle of the treated PLA fabric is measured to be approximately 150 degrees, which is high enough to exhibit the Lotus effect as a result of the superhydrophobicity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
175560
Triethoxyvinylsilane, 97%
C8H18O3Si
679275
Triethoxyvinylsilane, ≥98%, deposition grade
C8H18O3Si