EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Synthesis of an azide-tagged library of 2,3-dihydro-4-quinolones.
PMID 20141224
Abstract
We describe the assembly of a 960-member library of tricyclic 2,3-dihydro-4-quinolones using a combination of solution-phase high-throughput organic synthesis and parallel chromatographic purification. The library was produced with high efficiency and complete chemo- and diastereoselectivity by diversification of an azide-bearing quinolone via a sequence of [4 + 2] cycloadditions, N-acylations, and reductive aminations. The azide-functionalization of this library is designed to facilitate subsequent preparation of fluorescent or affinity probes, as well as small-molecule/surface conjugation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
262536
trans-3-Nonen-2-one, 95%
C9H16O