EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Addendum to Fragrance material review on Nerolidol (isomer unspecified).
PMID 20141875
Abstract
An addendum to the toxicologic and dermatologic review of Nerolidol (isomer unspecified) when used as a fragrance ingredient is presented.