EMAIL THIS PAGE TO A FRIEND
Organic letters
Diastereo- and enantioselective synthesis of alpha,gamma-diaminobutyric acid derivatives via Cu-catalyzed asymmetric Michael reaction.
PMID 20143857
Abstract
The first highly diastereo- and enantioselective catalytic asymmetric Michael addition of glycine derivatives to nitroalkenes have been developed. The enantioselectivity of ortho-substituted products can be significantly improved by using a new 1,2-P,N-ferrocene ligand L5. The alpha,gamma-diaminoacid derivative was obtained without the loss of optical activity from the adduct.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32830
L-2,4-Diaminobutyric acid dihydrochloride, ≥95.0%
C4H10N2O2 · 2HCl