EMAIL THIS PAGE TO A FRIEND
Nano letters
Ta3N5 nanotube arrays for visible light water photoelectrolysis.
PMID 20146505
Abstract
Tantalum nitride (Ta3N5) has a band gap of approximately 2.07 eV, suitable for collecting more than 45% of the incident solar spectrum energy. We describe a simple method for scale fabrication of highly oriented Ta3N5 nanotube array films, by anodization of tantalum foil to achieve vertically oriented tantalum oxide nanotube arrays followed by a 700 degrees C ammonia anneal for sample crystallization and nitridation. The thin walled amorphous nanotube array structure enables transformation from tantalum oxide to Ta3N5 to occur at relatively low temperatures, while high-temperature annealing related structural aggregation that commonly occurs in particle films is avoided. In 1 M KOH solution, under AM 1.5 illumination with 0.5 V dc bias typical sample (nanotube length approximately 240 nm, wall thickness approximately 7 nm) visible light incident photon conversion efficiencies (IPCE) as high as 5.3% were obtained. The enhanced visible light activity in combination with the ordered one-dimensional nanoarchitecture makes Ta3N5 nanotube arrays films a promising candidate for visible light water photoelectrolysis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF34465888
Tantalum, foil, 0.5m coil, thickness 0.034mm, as rolled, 99.9%
Ta
GF96084603
Tantalum, foil, 0.5m coil, thickness 0.03mm, as rolled, 99.9%
Ta
GF78648092
Tantalum, foil, 0.5m coil, thickness 0.03mm, coil width 1.2mm, annealed, 99.9%
Ta
GF38365106
Tantalum, foil, 0.5m coil, thickness 0.04mm, coil width .65mm, as rolled, 99.9%
Ta
GF76540181
Tantalum, foil, 0.5m coil, thickness 0.04mm, coil width 0.7mm, annealed, 99.9%
Ta
GF08928108
Tantalum, foil, 0.5m coil, thickness 0.04mm, coil width 1mm, annealed, 99.9%
Ta
GF61023574
Tantalum, foil, 0.5m coil, thickness 0.05mm, annealed, 99.9%
Ta
GF07210347
Tantalum, foil, 10m coil, thickness 0.025mm, coil width 0.75mm, annealed, 99.9%
Ta
GF10417698
Tantalum, foil, 10m coil, thickness 0.03mm, coil width 0.75mm, annealed, 99.9%
Ta
GF53229269
Tantalum, foil, 10m coil, thickness 0.04mm, coil width .65mm, as rolled, 99.9%
Ta
GF76534500
Tantalum, foil, 100x100mm, thickness 0.100mm, annealed, 99.9%
Ta
GF80276182
Tantalum, foil, 100x100mm, thickness 0.125mm, annealed, 99.9%
Ta
GF85784653
Tantalum, foil, 100x100mm, thickness 0.15mm, annealed, 99.9%
Ta
GF65422739
Tantalum, foil, 100x100mm, thickness 0.15mm, annealed, flattened, 99.9%
Ta
GF86342485
Tantalum, foil, 100x100mm, thickness 0.20mm, annealed, 99.9%
Ta
GF60298195
Tantalum, foil, 100x100mm, thickness 0.25mm, annealed, 99.9%
Ta
GF32441543
Tantalum, foil, 100x100mm, thickness 0.25mm, annealed, 99.99%
Ta
GF48780142
Tantalum, foil, 100x100mm, thickness 0.5mm, annealed, 99.9%
Ta
GF66145260
Tantalum, foil, 100x100mm, thickness 0.5mm, annealed, polished on both sides, 99.9%
Ta
GF55289548
Tantalum, foil, 15mm disks, thickness 0.5mm, annealed, 99.9%
Ta
GF64539872
Tantalum, foil, 15mm disks, thickness 0.75mm, annealed, 99.9%
Ta
GF24066525
Tantalum, foil, 1m coil, thickness 0.006mm, 99.9%
Ta
GF50723243
Tantalum, foil, 1m coil, thickness 0.009mm, 99.9%
Ta
GF45003204
Tantalum, foil, 1m coil, thickness 0.015mm, 99.9%
Ta
GF94153754
Tantalum, foil, 1m coil, thickness 0.025mm, as rolled, 99.9%
Ta
GF19196310
Tantalum, foil, 1m coil, thickness 0.025mm, coil width 0.75mm, annealed, 99.9%
Ta
GF13236487
Tantalum, foil, 1m coil, thickness 0.03mm, coil width 0.75mm, annealed, 99.9%
Ta
GF53856186
Tantalum, foil, 1m coil, thickness 0.03mm, coil width 1.2mm, annealed, 99.9%
Ta
GF10672365
Tantalum, foil, 1m coil, thickness 0.04mm, coil width .65mm, as rolled, 99.9%
Ta
GF67523351
Tantalum, foil, 1m coil, thickness 0.04mm, coil width 0.7mm, annealed, 99.9%
Ta