EMAIL THIS PAGE TO A FRIEND
Neurogastroenterology and Motility
Localization of muscarinic receptors M1R, M2R and M3R in the human colon.
PMID 20146726
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB1529
Anti-Nitric Oxide Synthase I Antibody, serum, clone, Chemicon®