EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Excited state intramolecular proton transfer (ESIPT) in six-coordinated zinc(ii)-quinoxaline complexes with ligand hydrogen bonds: their fluorescent properties sensitive to axial positions.
PMID 20148216
Abstract
Zinc(ii)-quinoxaline complexes, [Zn(hqxc)(2)(py)(2)] and [Zn(hqxc)(2)(DMSO)(2)] (hqxc = 3-hydroxy-2-quinoxalinecarboxylate, py = pyridine, DMSO = dimethyl sulfoxide), were prepared and characterized by X-ray crystallography and fluorescence spectroscopy. In both complexes, the zinc ion is six-coordinated by two equatorial bidentate hqxc ligands with an intramolecular hydrogen bond and two axial monodentate ligands such as pyridine or DMSO. In spite of similar coordination geometries, there is a remarkable difference between their solid-state fluorescent properties. The pyridine complex is strongly fluorescent (fluorescence quantum yield Phi = 0.22), giving rise to a significantly Stokes-shifted spectrum. From its thin film photopumped by a nitrogen gas laser, amplified spontaneous emission was observed. These results suggest that the fluorescence occurs by way of excited-state intramolecular proton-transfer (ESIPT) in the hydrogen bond of hqxc. On the other hand, the DMSO complex shows fluorescent intensity (Phi = 0.08) lower than that of the pyridine complex, and shows normal emission in addition to ESIPT emission. From IR measurements for these complexes, it is concluded that axial ligands influence the hydrogen bond strength of the equatorial hqxc ligand via zinc and thus the ESIPT efficiency.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
108340
3-Hydroxy-2-quinoxalinecarboxylic acid, 97%
C9H6N2O3