EMAIL THIS PAGE TO A FRIEND
British journal of pharmacology
Binding of GTPgamma[35S] is regulated by GDP and receptor activation. Studies with the nociceptin/orphanin FQ receptor.
PMID 20148892
Abstract
We have examined the effects of ligand efficacy and receptor density on the binding of guanosine 5'-[gamma-thio]triphosphate (GTPgammaS) and GDP to the nociceptin/orphanin FQ (N/OFQ) peptide receptor (NOP)-coupled G-proteins. In GTPgamma[(35)S] binding experiments, using stable (CHO(hNOP)) and inducible (CHO(INDhNOP)) recombinant human and rat NOP we have measured: (i) ligand-specific GDP requirements; (ii) the effects of receptor density on guanine nucleotide affinity/capacity; and (iii) the effect of ligand efficacy on GTPgammaS association kinetics. GTPgammaS competition curves were shallow and modelled by high- and low-affinity components that were relatively consistent between cell types and tissue preparations. In the presence of 1 microM N/OFQ a high-affinity GDP binding site was also present, but the fraction of total binding was reduced. In an efficacy-dependent manner, the partial agonists [F/G]N/OFQ(1-13)NH(2) ([Phe(1)psi(CH(2)-NH)Gly(2)]-nociceptin(1-13)NH(2)) and naloxone benzoylhydrazone both reduced the fraction of high-affinity sites for GDP (relative to basal). While the pIC(50) for high-affinity GDP binding site did not decrease in the presence of 1 microM N/OFQ, N/OFQ produced a significant reduction in pIC(50) for the low-affinity site. Agonist-mediated decrease in affinity for GDP binding was efficacy-dependent. GDP displayed three affinities: high, conserved in the presence and absence of ligand; intermediate, present as a low fraction under basal conditions; low (efficacy-dependent), present during receptor activation representing the majority of binding. The affinity of GTPgamma[(35)S] was regulated by GDP and receptor activation caused increased binding of GTPgamma[(35)S] through a reduction in GDP affinity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N165
Naloxone benzoylhydrazone, solid
C26H27N3O4
P3490
Ponasterone A, ≥65%
C27H44O6