EMAIL THIS PAGE TO A FRIEND
Pest management science
Suppression of pine wilt disease by an antibacterial agent, oxolinic acid.
PMID 20151406
Abstract
Pine wilt disease (PWD) is very complex and has been reported to be caused by pine wood nematode, Bursaphelenchus xylophilus (Steiner & Buhrer) Nickle, and its accompanying bacteria. However, there is no report on the control of PWD by antibacterial agent. The present study was performed to investigate disease control efficacy of antibacterial agents against PWD. Among six antibacterial antibiotics tested, oxolinic acid (OA) showed the strongest antibacterial activity against five bacteria isolated from three strains of pine wood nematode. In in vivo assay, it effectively suppressed the development of PWD in three-year-old seedlings of Pinus densiflora Sieb. & Zucc.; it showed 71% control when injected at 3 mg per seedling. A mixture of OA and the nematicidal agent abamectin (Ab) showed higher disease control efficacy against PWD than either OA or Ab alone. In addition, OA alone and a mixture of OA and Ab also controlled PWD in approximately 20-year-old pine trees under field conditions. This is the first report on the suppression of PWD by OA. The result strongly indicates that PWD could be controlled by antibacterial antibiotic alone and a combination of antibacterial and nematicidal agents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O0877
Oxolinic acid, quinolone antibiotic
C13H11NO5
67126
Oxolinic acid, analytical standard
C13H11NO5