EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
The mechanism of nucleophilic displacements at phosphorus in chloro-substituted methylphosphonate esters: P-O vs P-C bond cleavage: a DFT study.
PMID 20151695
Abstract
Potential energy surfaces for the nucleophilic displacements at phosphorus in dimethyl methyl-, chloromethyl-, dichloromethyl-, and trichloromethylphosphonates have been computed at the B3LYP/6-31+G* level of theory, using IEF-PCM to account for the solvent effect. The results reveal that sequential addition of chlorine substituents on the methyl phosphonates increases the stability of transition states and intermediates which facilitate P-C bond cleavage. Thus, while nonsubstituted dimethyl methylphosphonate and dimethyl chloromethylphosphonate may undergo exclusive P-O bond cleavage, the trichlorinated analogue exclusively undergoes P-C bond dissociation. Dichloromethylphosphonic acid derivatives were found to be borderline cases: while P-O fission is the preferred process, P-C scission might also be feasible. The increase in stability of the corresponding transition states and intermediates can account for the enhancement in the apicophilicity of the methyl ligand upon substitution with chlorine atoms.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
289868
Methylphosphonic acid, 98%
CH5O3P