EMAIL THIS PAGE TO A FRIEND
BJU international
The dapoxetine paradox.
PMID 20151958
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D7071
Dapoxetine hydrochloride, ≥98% (HPLC)
C21H23NO · HCl