EMAIL THIS PAGE TO A FRIEND
Behavioural brain research
Spatial memory deficits and thalamic serotonergic metabolite change in thiamine deficient rats.
PMID 20153779
Abstract
The purposes of the present study were to verify the effects of a severe thiamine deficiency episode on spatial cognitive aspects and thalamic serotonergic parameters. The animals were submitted to a severe thiamine deficiency treatment that was interrupted after the onset of the last neurological signs. The results obtained confirm previous findings about TD deficiency effects on cognitive function and, further show that this vitamin increases the thalamic serotonine metabolite, 5-hidroxyindolacetic acid (5-HIAA), level. In addition, the present data shed light on the importance of this metabolite in spatial cognitive function.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0256
Pyrithiamine hydrobromide, ~95%, crystalline
C14H18N4O · 2HBr