EMAIL THIS PAGE TO A FRIEND
Electrophoresis
Achiral and chiral separations using MEKC, polyelectrolyte multilayer coatings, and mixed mode separation techniques with molecular micelles.
PMID 20155738
Abstract
Mixed mode (MM) separation using a combination of MEKC and polyelectrolyte multilayer (PEM) coatings is herein reported for the separation of achiral and chiral analytes. Many analytes are difficult to separate by MEKC and PEM coatings alone. Therefore, the implementation of a MM separation provides several advantages for overcoming the limitations of these well-established methods. In this study, it was observed that achiral separations using MEKC and PEM coatings individually resulted in partial resolution of eight very similar aryl ketones when the molecular micelle (sodium poly(N-undecanoyl-L-glycinate)) concentration was varied from 0.25 to 1.00% w/v and the bilayer number varied from 2 to 4. However, when MM separation was introduced, baseline resolution was achieved for all eight analytes. In the case of chiral separations, temazepam, aminoglutethimide, benzoin, benzoin methyl ether, and coumachlor were separated using the three separation techniques. For chiral separations, the chiral molecular micelle, sodium poly(N-undecanoyl-L-leucylvalinate), was employed at concentrations of 0.25-1.50% w/v for both MEKC and PEM coatings. Overall, the results revealed partial separation with MEKC and PEM coatings individually. However, MM separation enabled baseline separation of each chiral mixture. The separation of achiral and chiral compounds from different compound classes demonstrates the versatility of this MM approach.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
189219
Coumachlor, 98%
C19H15ClO4
A9657
DL-Aminoglutethimide
C13H16N2O2
T8275
Temazepam, analytical standard
C16H13ClN2O2