EMAIL THIS PAGE TO A FRIEND
Organic letters
Direct and selective functionalization of a tetrahydro-beta-carboline at position 4.
PMID 20158220
Abstract
The first example of direct functionalization of a tetrahydro-beta-carboline (THBC) at position 4 is presented. Nucleophiles with various reactivity add to a transient intermediate generated in situ from this THBC. This reaction proceeds with a total control of the two created stereogenic centers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
300764
1,2,3,4-Tetrahydro-9H-pyrido[3,4-b]indole, 98%
C11H12N2