EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Botulinum neurotoxin A protease: discovery of natural product exosite inhibitors.
PMID 20158239
Abstract
A new mechanistic class of BoNT/A zinc metalloprotease inhibitors, from Echinacea, exemplified by the natural product d-chicoric acid (I1) is disclosed. A detailed evaluation of chicoric acid's mechanism of inhibition reveals that the inhibitor binds to an exosite, displays noncompetitive partial inhibition, and is synergistic with a competitive active site inhibitor when used in combination. Other components found in Echinacea, I3 and I4, were also inhibitors of the protease.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06957
( −)-Chicoric acid, analytical standard
C22H18O12