EMAIL THIS PAGE TO A FRIEND
Pediatrics international : official journal of the Japan Pediatric Society
"Typhlitis" in rectum.
PMID 20158643
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N0755 Netilmicin sulfate salt
C21H41N5O7 · 2.5H2O4S